Learn More
New Pig™ HazMat Spill Kits
Product Code. 12913625
Encompass_Preferred
Change view
Click to view available options
Capacity (Metric):
13.62 L
34.068 L
79.49 L
For Use With (Application):
Acids, Bases, Unknowns
Acids, Bases, Unknowns Fluid Absorption
Oils, Coolants, Solvents, Water
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) For Use With (Application) Unit Size
12913625 79.49 L Oils, Coolants, Solvents, Water Each
12903595 13.62 L Acids, Bases, Unknowns Fluid Absorption Each
12963615 34.068 L Oils, Coolants, Solvents, Water Each
12913585 34.068 L Acids, Bases, Unknowns Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12913625 Supplier New Pig™ Supplier No. RFL236
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Durable, water-resistant bag with hook-and-loop closure keeps contents clean and dry.
  • Feature PIG absorbents proven for rapid response
  • Socks stop spreading spills; PIG HazMat Pads absorb quickly
  • Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
  • Temporary disposal bags help make cleanup easier
  • Bright yellow bag and large, color-coded label make the kit easy to locate
  • Strap for easy carrying and hanging
  • Compact and easy to store
Specifications
Capacity (Metric) 79.49 L
For Use With (Application) Oils, Coolants, Solvents, Water