missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HB-EGF Antibody (125923), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB259T
This item is not returnable.
View return policy
Description
HB-EGF Monoclonal antibody specifically detects HB-EGF in Human samples. It is validated for Western Blot
Specifications
| HB-EGF | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 125923 | |
| Western Blot | |
| diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), DTS, DTSF, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HB-EGF, Asp63-Leu148, Accession # Q53H93 | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 1839 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction