missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HBAE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143417
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. Positive control: zebrafish heart, zebrafish muscle, whole zebrafish.
Specifications
| HBAE1 | |
| Polyclonal | |
| Unconjugated | |
| hbae1 | |
| ae1; alphae1globin; dZ118J2.6; embryonic alpha globin e1; etID40432.3; globin alphae1; HbA; hba2; HBA2r; HBAA1; hbae1; hbae1.1; hemoglobin alpha embryonic-1; hemoglobin alpha-2 chain; hemoglobin, alpha embryonic 1; hemoglobin, alpha embryonic 1.1; si:by119c3.1; si:by119c3.3; SI:dZ118J2.10; SI:dZ118J2.8; si:xx-119c3.3; wu:fa12a11; wu:fb74f06; zgc:86936 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 30597 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 0.54 mg/mL | |
| PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
| 0 | |
| hbae1 | |
| Recombinant protein encompassing a sequence within the center region of zebrafish Hbae1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction