missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HBB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560287
This item is not returnable.
View return policy
Description
Immunogen sequence: FATLSELHCD KLHVDPENFR LLGN Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
The alpha (HBA) and beta (HBB) loci determine the structure of the 2 types of polypeptide chains in adult hemoglobin, Hb A. The normal adult hemoglobin tetramer consists of two alpha chains and two beta chains. Mutant beta globin causes sickle cell anemia. Absence of beta chain causes beta-zero-thalassemia. Reduced amounts of detectable beta globin causes beta-plus-thalassemia. The order of the genes in the beta-globin cluster is 5'-epsilon-gamma-G-gamma-A-delta-beta-3'.
Specifications
| HBB | |
| Polyclonal | |
| Unconjugated | |
| HBB | |
| beta globin chain; beta-1-globin; Beta-globin; CD113t-C; Hbb; hemoglobin beta; Hemoglobin beta chain; hemoglobin beta chain complex; hemoglobin beta chain, major-form; Hemoglobin beta-1 chain; hemoglobin subunit beta; Hemoglobin subunit beta-1; hemoglobin, beta; III beta-1 globin; III beta-2 globin; LVV-hemorphin-7; Spinorphin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3043 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P68871 | |
| HBB | |
| Recombinant protein corresponding to Human HBB. Recombinant protein control fragment (Product #RP-100088). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction