missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ HBC 530
GREENER_CHOICE

Product Code. 18769286
Click to view available options
Quantity:
60 μg
Unit Size:
60µg
This item is not returnable. View return policy

Product Code. 18769286

Brand: Tocris Bioscience™ 7277/60U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GFP fluorophore mimic for imaging RNA in live cells; activated by binding to Pepper aptamers; suitable for confocal and two-photon microscopy

HBC 530 is a mimic of green fluorescent protein (GFP) fluorophore for imaging RNA in live cells. Fluoresces when bound to Pepper aptamer; displays high affinity binding (Kd = 3.5 nM). Suitable for confocal and two-photon microscopy. Confocal excitation maximum = 488 nm, emission maximum= 495 - 595 nm. Two-photon excitation maximum = 976 nm, emission maximum= 475 - 575 nm. To make a 1000x stock solution, add 200 μL DMSO (Cat. No. 3176).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Cyano-α-[[4-[(2-hydroxyethyl)methylamino]phenyl]methylene]benzeneacetonitrile
CAS 156840-13-0
Quantity 60 μg
Target Fluorescent RNA Probes
Molecular Formula C19H17N3O
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.