missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBcAg Antibody (H3A4cc) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB110-2512JF585
This item is not returnable.
View return policy
Description
HBcAg Monoclonal antibody specifically detects HBcAg in Virus samples. It is validated for ELISA,Western Blot
Specifications
| HBcAg | |
| Monoclonal | |
| Janelia Fluor 585 | |
| c, Core antigen, HBc, HBcAg, Hepatitis B Virus core antigen | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant HBcAg. | |
| 0.2 mL | |
| Virology Bacteria and Parasites | |
| 944568 | |
| Store at 4°C in the dark. | |
| IgG2a |
| ELISA, Western Blot | |
| H3A4cc | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction