Learn More
HBD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16824-1-AP-20UL
This item is not returnable.
View return policy
Description
Hemoglobin subunit delta is a protein that in humans is encoded by the HBD gene. The delta (HBD) and beta (HBB) genes are normally expressed in the adult: two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin. Two alpha chains plus two delta chains constitute HbA-2, which with HbF comprises the remaining 3% of adult hemoglobin. Five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5′-epsilon-Ggamma-Agamma-delta-beta-3′.Specifications
| HBD | |
| Polyclonal | |
| Unconjugated | |
| HBD | |
| Delta globin, HBD, Hemoglobin delta chain, Hemoglobin subunit delta, hemoglobin, delta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100187828 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02042 | |
| HBD | |
| HBD Fusion Protein Ag10409 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |