Learn More
HBD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16824-1-AP-20UL
 View more versions of this product
Product Code. 16896574
  • € 145.00 / 20µL
Description
Hemoglobin subunit delta is a protein that in humans is encoded by the HBD gene. The delta (HBD) and beta (HBB) genes are normally expressed in the adult: two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin. Two alpha chains plus two delta chains constitute HbA-2, which with HbF comprises the remaining 3% of adult hemoglobin. Five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5′-epsilon-Ggamma-Agamma-delta-beta-3′.
Specifications
Specifications
HBD
Polyclonal
Unconjugated
HBD
Delta globin, HBD, Hemoglobin delta chain, Hemoglobin subunit delta, hemoglobin, delta
Rabbit
Antigen Affinity Chromatography
RUO
100187828
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02042
HBD
HBD Fusion Protein Ag10409
20 μL
Primary
Human, Rat, Mouse