missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBEGF Monoclonal Antibody (OTI6F4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812466
This item is not returnable.
View return policy
Description
HBEGF is a growth factor that mediates its effects via EGFR, ERBB2 and ERBB4. It is required for normal cardiac valve formation and normal heart function. HBEGF promotes smooth muscle cell proliferation. It may be involved in macrophage-mediated cellular proliferation. It is mitogenic for fibroblasts, but not endothelial cells. It is able to bind EGF receptor/EGFR with higher affinity than EGF itself and is a far more potent mitogen for smooth muscle cells than EGF. It also acts as a diphtheria toxin receptor.Specifications
| HBEGF | |
| Monoclonal | |
| Unconjugated | |
| HBEGF | |
| DTR, DTS, DTSF, HEGFL | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1839 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI6F4 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99075 | |
| HBEGF | |
| Human recombit protein fragment corresponding to amino acids 20-208 of human HBEGF produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction