missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBEGF Monoclonal Antibody (OTI8B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812468S
This item is not returnable.
View return policy
Description
HBEGF Monoclonal antibody specifically detects HBEGF in Human samples. It is validated for Western BlotSpecifications
| HBEGF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99075 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1839 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8B5 | |
| Unconjugated | |
| HBEGF | |
| AW047313; Diphtheria toxin receptor; Diphtheria toxin receptor (heparin binding epidermal growth factor - like growth factor); diphtheria toxin receptor (heparin-binding EGF-like growth factor); diphtheria toxin receptor (heparin-binding epidermal growth factor-like growth factor); DTR; DT-R; DTS; DTSF; GFHB; hb egf; HBEGF; Hb-egf; Hegfl; heparin binding EGF like growth factor; heparin binding epidermal growth factor-like growth factor; heparin-binding EGF-like growth factor; heparin-binding epidermal -growth - like growth factor; heparin-binding epidermal growth factor; heparin-binding epidermal -growth factor; proheparin-binding EGF-like growth factor | |
| Human recombinant protein fragment corresponding to amino acids 20-208 of human HBEGF produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction