missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBS1L Monoclonal Antibody (OTI1F11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800566
This item is not returnable.
View return policy
Description
This gene encodes a member of the GTP-binding elongation factor family. It is expressed in multiple tissues with the highest expression in heart and skeletal muscle. The intergenic region of this gene and the MYB gene has been identified to be a quantitative trait locus (QTL) controlling fetal hemoglobin level, and this region influnces erythrocyte, platelet, and monocyte counts as well as erythrocyte volume and hemoglobin content. DNA polymorphisms at this region associate with fetal hemoglobin levels and pain crises in sickle cell disease. A single nucleotide polymorphism in exon 1 of this gene is significantly associated with severity in beta-thalassemia/Hemoglobin E. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene.Specifications
| HBS1L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y450 | |
| HBS1L | |
| Human recombit protein fragment corresponding to amino acids 1-260 of human HBS1L produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1F11 | |
| Unconjugated | |
| HBS1L | |
| EF-1a, ERFS, HBS1, HSPC276, eRF3c | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10767 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction