missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBS1L Monoclonal Antibody (OTI1G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800608S
This item is not returnable.
View return policy
Description
HBS1L Monoclonal antibody specifically detects HBS1L in Human samples. It is validated for Western BlotSpecifications
| HBS1L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y450 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10767 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1G7 | |
| Unconjugated | |
| HBS1L | |
| 2810035F15Rik; AI326327; EF-1a; eRF3 family member; eRF3c; ERF3-similar protein; ERFS; eukaryotic release factor 3; Hbs1; HBS1 like translational GTPase; HBS1L; Hbs1-like (S. cerevisiae); HBS1-like protein; HBS1-like translational GTPase; Hsp70 subfamily B suppressor 1-like protein; HSPC276; Kiaa1038 | |
| Human recombinant protein fragment corresponding to amino acids 1-242 of human HBS1L produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction