missing translation for 'onlineSavingsMsg'
Learn More

HBsAg Antibody (5C3), Novus Biologicals™

Product Code. p-200132262 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
1 mL
Unit Size:
0.1mL
0.5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30532825 0.1 mL 0.1mL
30532826 0.5 mL 0.5mL
30532827 1 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30532825 Supplier Novus Biologicals Supplier No. NBP3451130.1ml

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HBsAg Monoclonal antibody specifically detects HBsAg in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HBsAg
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5C3
Conjugate Unconjugated
Formulation PBS (pH 7.4), Tween-20, 0.2% BSA
Gene Alias HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S
Host Species Mouse
Immunogen HBsAg (ad/ay) protein
Purification Method Tissue culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 944569
Target Species Human
Content And Storage Store at 4°C.
Form Supernatant
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.