missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBsAg Antibody (HB11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-7391H
This item is not returnable.
View return policy
Description
HBsAg Monoclonal specifically detects HBsAg in Virus samples. It is validated for ELISA, Sandwich ELISA.
Specifications
| HBsAg | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| S | |
| Recombinant full length protein (HBsAg of ayw subtype). | |
| 0.2 mL | |
| Virology Bacteria and Parasites | |
| 944569 | |
| Virus | |
| Purified |
| ELISA | |
| HB11 | |
| ELISA, Sandwich ELISA | |
| HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reacts with the following HbsAg subtypes : ayw1, ayw2, ayw3, ayw4, ayr, adw2, adw4, adrq+, adrq-, ayw3 (Fer). | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction