missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBsAg Antibody (HB5), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-475JF646
This item is not returnable.
View return policy
Description
HBsAg Monoclonal specifically detects HBsAg in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| HBsAg | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| S | |
| purified HbsAg from human plasma | |
| 0.1 mL | |
| Primary | |
| This antibody (clone HB5) recognizes following Hepatitis B virus (HBV) subtypes: ayw2, ayw3, ayw4, ayr, adw2, adw4, adrq+, adrq-. Hepatitis B surface antigen (HBsAg) is a marker of infectivity. Specificity of the antibody HB5 was verified by ELISA on panel of virus subtypes identified on International Workshop on HBsAg Subtypes (Paris, April 1975). The antibody HB5 does not cross-block with This antibody HB3. | |
| Store at 4°C in the dark. | |
| IgG2a |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| HB5 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
| Mouse | |
| Ion exchange chromatography | |
| Cell Biology, Virology Bacteria and Parasites | |
| 944569 | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction