missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies HBSS, Modified (Without Ca++ and Mg++)

Product Code. 17198091
Encompass_Preferred
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 17198091

Brand: STEMCELL Technologies 37250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hanks' Balanced Salt Solution (HBSS) without calcium and magnesium

  • Hanks' Balanced Salt Solution (HBSS) can be used for diluting, washing, irrigating, or as a transporting solution for cell or tissue culture
  • It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media, and provides cells with a source of water and essential inorganic ions, and a carbohydrate as an energy source
  • Selection of suitable nutrient medium is dependent on type of cell, culture conditions, and degree of chemical definition required for the cell culture application

Specifications

Quantity 500 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.