missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCFC1R1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563761
This item is not returnable.
View return policy
Description
Immunogen sequence: QPLQRGPQGG AQRLPRAALG VTWGLDASSP LRGAVPMSTK RRLEEEQEPL RKQFLSEEN Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 46%.
Regulates HCFC1 activity by modulating its subcellular localization. Overexpression of HCFC1R1 leads to accumulation of HCFC1 in the cytoplasm. HCFC1R1-mediated export may provide the pool of cytoplasmic HCFC1 required for import of virion-derived VP16 into the nucleus.
Specifications
| HCFC1R1 | |
| Polyclonal | |
| Unconjugated | |
| HCFC1R1 | |
| HCF-1 beta-propeller-interacting protein; HCFC1R1; Host cell factor C1 regulator 1; host cell factor C1 regulator 1 (XPO1 dependent); HPIP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54985 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NWW0 | |
| HCFC1R1 | |
| Recombinant protein corresponding to Human HCFC1R1. Recombinant protein control fragment (Product #RP-101727). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction