missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCFC2 Monoclonal Antibody (OTI6C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505236
This item is not returnable.
View return policy
Description
This gene encodes one of two proteins which interact with VP16, a herpes simplex virus protein that initiates virus infection. Both the encoded protein and the original Herpes host cell factor interact with VP16 through a beta-propeller domain. The original Herpes host cell factor, however, is effective at initiating viral infection while the encoded protein is not. Transcripts of varying length due to alternative polyadenylation signals have been described.Specifications
| HCFC2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y5Z7 | |
| HCFC2 | |
| Full length human recombit protein of human HCFC2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI6C9 | |
| Unconjugated | |
| HCFC2 | |
| HCF-2, HCF2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29915 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction