missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCFC2 Mouse anti-Human, Clone: OTI5H9, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505235S
This item is not returnable.
View return policy
Description
HCFC2 Monoclonal antibody specifically detects HCFC2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| HCFC2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y5Z7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29915 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI5H9 | |
| Unconjugated | |
| HCFC2 | |
| 1700129L13Rik; AU019838; C2 factor; HCF2; HCF-2; HCFC2; Host cell factor 2; host cell factor C2 | |
| Full length human recombinant protein of human HCFC2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction