missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144762
This item is not returnable.
View return policy
Description
Immunogen sequence: CMKSKFLQVG GNTFSKTETS ASPHCPVYVP DPTSTIKPGP NSHNSNTPGI REAGSEDIIV VA Antigen sequence identity: Mouse 61%; Rat 60%.
Hematopoietic cell kinase (HCK) is a SRC-related kinase expressed in lymphoid and hematopoietic cells and may play a role in some forms of leukemia. (HCK Alias: DKFZp686D0638). HCK is a protein-tyrosine kinase that is predominantly expressed in hemopoietic cell types. The encoded protein may help couple the Fc receptor to the activation of the respiratory burst. In addition, it may play a role in neutrophil migration and in the degranulation of neutrophils. Alternate translation initiation site usage, including a non-AUG (CUG) codon, results in the production of two different isoforms, that have different subcellular localization.
Specifications
| HCK | |
| Polyclonal | |
| Unconjugated | |
| HCK | |
| AI849071; B cell/myeloid kinase; B-cell/myeloid kinase; BMK; HCK; HCK proto-oncogene, Src family tyrosine kinase; Hctk; Hematopoietic cell kinase; hemopoietic cell kinase; hemopoietic cell tyrosine kinase; JTK9; p56Hck; p56-HCK; p56-HCK/p59-HCK; p59Hck; p59-HCK/p60-HCK; p61Hck; tyrosine-protein kinase HCK | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3055 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P08631 | |
| HCK | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 24-85. Recombinant protein control fragment (Product #RP-109909). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction