missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX Hck Protein, Active

Product Code. 17320374
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17320374 10 μg 10µg
17330374 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17320374 Supplier Eurofins DiscoverX Supplier No. 14577

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal His6-tagged, recombinant, human Hck, amino acids 230-497, for use in Kinase Assays.

Hematopoietic cell kinase (HCK) is a member of Src protein-tyrosine kinase family and expressed primary in granulocytes, monocytes and macrophages. Active HCK phosphorylates STAT5B on Tyr699, which represents an essential step in Signal transducer and activator of transcription 5 (STAT5B) stimulation. The binding of SH3 ligands, such as the Nef protein of HIV-1, further augments Hck activity suggesting a new mechanism for regulating tyrosine kinase activity.

GenBank NM_002110

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 34.1 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥59%
Protein Hck, active
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.