missing translation for 'onlineSavingsMsg'
Learn More

HCN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-35653-20ul

 View more versions of this product

Product Code. 30229790

  • € 190.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HCN1 Polyclonal antibody specifically detects HCN1 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HCN1
Polyclonal
Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
BCNG1, BCNG-1brain cyclic nucleotide gated channel 1, Brain cyclic nucleotide-gated channel 1, HAC-2, hyperpolarization activated cyclic nucleotide-gated potassium channel 1, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1
A synthetic peptide corresponding to a sequence within amino acids 791-890 of human HCN1 (NP_066550.2).,, Sequence:, LISRPHPTVGESLASIPQPVTAVPGTGLQAGGRSTVPQRVTLFRQMSSGAIPPNRGVPPAPPPPAAALPRESSSVLNTDPDAEKPRFASNL
20 μL
Cardiovascular Biology, Neuroscience
348980
Store at -20°C. Avoid freeze-thaw cycles.
IgG
ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.