missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14084
This item is not returnable.
View return policy
Description
HCN1 Polyclonal antibody specifically detects HCN1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| HCN1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| BCNG1, BCNG-1brain cyclic nucleotide gated channel 1, Brain cyclic nucleotide-gated channel 1, HAC-2, hyperpolarization activated cyclic nucleotide-gated potassium channel 1, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: PSLPHEVSTLISRPHPTVGESLASIPQPVTAVPGTGLQAGGRSTVPQRVTLFRQMSSGAIPPNRGVPPAPPPPAAALPRESSSVLNTDPDAE | |
| 0.1 mL | |
| Cardiovascular Biology, Neuroscience | |
| 348980 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?