Learn More
HCN2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30946-1-AP-20UL
 View more versions of this product
Product Code. 18010636
  • € 145.00 / 20µL
Description
The HCN2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human HCN2. This antibody recognizes human, rat, mouse antigen. The HCN2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
HCN2
400 μg/mL
Rabbit
Primary
Human, Rat, Mouse
150 kDa
IgG
Western Blot, ELISA
HAC 1, HCN2
20 μL
610
Store at -20°C. Stable for one year after shipment.