Learn More
HCN2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30946-1-AP-150UL
This item is not returnable.
View return policy
Description
The HCN2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human HCN2. This antibody recognizes human, rat, mouse antigen. The HCN2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| HCN2 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| HAC 1, HCN2 | |
| 150 μL | |
| 610 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |