missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCN3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142593
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
HCN3 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. HCN3 has been detected in the brain, but with less expression than other HCN channels.
Specifications
| HCN3 | |
| Polyclonal | |
| Unconjugated | |
| Hcn3 | |
| Bcng4; BCNG-4; Hac3; HAC-3; HCN3; Hcn4; hyperpolarization activated cyclic nucleotide gated potassium channel 3; hyperpolarization activated cyclic nucleotide-gated potassium channel 3; hyperpolarization-activated cation channel 3; hyperpolarization-activated cyclic nucleotide-gated potassium channel 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 4; KIAA1535; MGC131493; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 114245 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9JKA8 | |
| Hcn3 | |
| Peptide with sequence C-RGVRGRAPSTGAQ. | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction