missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCN3 Polyclonal Antibody

Description
Immunogen sequence: LPLSPDSPAT LLARSAWRSA GSPASPLVPV RAGPWASTSR LPAPPARTLH ASLSRAGRSQ VSLLGPPPGG GGRRLGPRGR PLSASQPSLP QRATGDGSPG RKGSGSERLP PSGLLAKPPR TAQPPRPPVP EPATPRGLQL S Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 88%.
HCN3 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. HCN3 has been detected in the brain, but with less expression than other HCN channels.
Specifications
Specifications
| Antigen | HCN3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Hcn3 |
| Gene Accession No. | Q9P1Z3 |
| Gene Alias | Bcng4; BCNG-4; Hac3; HAC-3; HCN3; Hcn4; hyperpolarization activated cyclic nucleotide gated potassium channel 3; hyperpolarization activated cyclic nucleotide-gated potassium channel 3; hyperpolarization-activated cation channel 3; hyperpolarization-activated cyclic nucleotide-gated potassium channel 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 4; KIAA1535; MGC131493; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 3 |
| Gene Symbols | Hcn3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?