missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCN4 Antibody (S114/10), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12910V
This item is not returnable.
View return policy
Description
HCN4 Monoclonal specifically detects HCN4 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| HCN4 | |
| Monoclonal | |
| DyLight 405 | |
| hyperpolarization activated cyclic nucleotide-gated cation channel 4, hyperpolarization activated cyclic nucleotide-gated potassium channel 4, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4, SSS2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 10021 | |
| Human, Mouse, Rat, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| S114/10 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| HCN4 | |
| Fusion protein amino acids 1-019-1198 (c-terminus) of rat HCN4 | |
| 0.1 mL | |
| Primary | |
| Detects approx 130 kDa. No cross-reactivity against other HCNs. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction