missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCV-NS5b Antibody (5B-3B1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80123
This item is not returnable.
View return policy
Description
HCV-NS5b Monoclonal specifically detects HCV-NS5b in Virus samples. It is validated for Western Blot.
Specifications
| HCV-NS5b | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot | |
| EC:2.7.7.48, Nonstructural Protein 5B, NS5B, Q8JXY1, RNA-directed RNA polymerase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes HCV-NS5b (epitope mapped to NS5B aa 372-382). It recognizes genotype 1 a and also 1b (albeit with lower affinity). It does not recognize genotype 2a. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot | |
| 5B-3B1 | |
| Unconjugated | |
| PBS containing 10% glycerol | |
| POLY | |
| Recombinant HCV-NS5b RNA-dependent RNA polymerase (genotype 1a). | |
| 100 μg | |
| Virology Bacteria and Parasites | |
| 951475 | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction