missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC1 Antibody (S09-7G9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19652-50ul
This item is not returnable.
View return policy
Description
HDAC1 Monoclonal antibody specifically detects HDAC1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
Specifications
| HDAC1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA | |
| S09-7G9 | |
| Unconjugated | |
| EC 3.5.1.98, GON-10, HD1DKFZp686H12203, histone deacetylase 1, reduced potassium dependency, yeast homolog-like 1, RPD3L1RPD3 | |
| A synthetic peptide of human HDAC1 (Uniprot # Q13547) | |
| 50 μg | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Loading Controls, Phospho Specific, Stem Cell Markers | |
| 3065 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction