missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HDAC1 Monoclonal Antibody (2A1D3)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA548488

Product Code. 17946061

  • € 512.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HDAC1 Monoclonal Antibody for ELISA, Flow, ICC/IF, IHC (P), Western Blot

Histone acetylation and deacetylation, catalyzed by multisubunit complexes, play a key role in the regulation of eukaryotic gene expression. HDAC1 belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex. It also interacts with retinoblastoma tumor-suppressor protein and this complex is a key element in the control of cell proliferation and differentiation. Together with metastasis-associated protein-2, HDAC1 deacetylates p53 and modulates its effect on cell growth and apoptosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HDAC1
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
O09106, Q13547
HDAC1
Purified recombinant fragment of human HDAC1 (AA: 321-482) expressed in E. Coli.
100 μg
Primary
Human, Mouse
Antibody
IgG1
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
2A1D3
Unconjugated
HDAC1
DKFZp686H12203; GON-10; HD1; Hdac1; Hdac1-ps; Histone deacetylase 1; histone deacetylase 1-like protein; MommeD5; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2; RPD3; RPD3L1
Mouse
Protein G
RUO
3065, 433759
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.