missing translation for 'onlineSavingsMsg'
Learn More

HDAC1 Polyclonal Antibody, Alexa Fluor™ 488, Invitrogen™

Product Code. 15957284
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15957284 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15957284 Supplier Invitrogen Supplier No. PA1860A488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1860A488 detects histone deacetylase 1 (HDAC1) from human, mouse, rat, hamster and canine tissues and cells. The PA1860A488 immunogen is a synthetic peptide corresponding to residues C E(467) E K P E A K G V K E E V K L A(482) of human HDAC1. This immunizing peptide (Cat. No. PEP-069) is available for use in neutralization and control experiments.

Chromatin is a highly specialized structure composed of tightly compacted chromosomal DNA. Gene expression within the nucleus is controlled, in part, by a host of protein complexes which continuously pack and unpack the chromosomal DNA. One of the known mechanisms of this packing and unpacking process involves the acetylation and deacetylation of the histone proteins comprising the nucleosomal core. Acetylated histone proteins confer accessibility of the DNA template to the transcriptional machinery for expression. Histone deacetylases (HDACs) are chromatin remodeling factors that deacetylate histone proteins and thus, may act as transcriptional repressors. HDACs are classified by their sequence homology to the yeast HDACs and there are currently 2 classes. Class I proteins are related to Rpd3 and members of class II resemble Hda1p. HDAC1 is a class I histone deacetylase containing 482 amino acid residues. HDAC1 has been shown to interact directly with transcription factors and has been shown to deacetylate histone proteins H3 and H4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene HDAC1
Gene Accession No. Q13547, O09106, Q4QQW4
Gene Alias HD1, RPD3L1, GON-10, RP4-811H24.2, RPD3, MommeD5, Hdac1-ps
Gene Symbols HDAC1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C E(467) E K P E A K G V K E E V K L A(482) of human HDAC1
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100752721, 297893, 3065, 433759, 487309
Target Species Canine, Hamster, Human, Mouse, Rat
Content And Storage 4° C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.