missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HDAC1 Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
:
0.1 mL
Specifications
Specifications
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1 M Urea, pH 7.4. |
| Gene ID (Entrez) | 3065 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 25 kDa |
| Quantity | 0.1 mL |
| Storage Requirements | Store at -20°C. Avoid freeze/thaw cycles. |
| Gene Alias | EC 3.5.1.98, GON-10, HD1DKFZp686H12203, histone deacetylase 1, reduced potassium dependency, yeast homolog-like 1, RPD3L1RPD3 |
| Gene Symbol | HDAC1 |
| Inhibitor Properties | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-52937. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL |
| Research Category | Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Loading Controls, Stem Cell Markers |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction