Learn More
Invitrogen™ HDAC10 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA599170
Product Code. 16603425
  • € 491.00 / 100µL
Description
Antibody detects endogenous levels of total HDAC10.
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HDAC10
Polyclonal
Unconjugated
HDAC10
AW548891; DKFZp761B039; Hd10; Hdac10; Histone deacetylase 10; MGC149722; Polyamine deacetylase HDAC10; RP3-402G11.7
Rabbit
Affinity chromatography
RUO
170787, 362981, 83933
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q569C4, Q6P3E7, Q969S8
HDAC10
A synthesized peptide derived from human HDAC10(Accession Q969S8), corresponding to amino acid residues A4-E54.
100 μL
Primary
Human, Mouse, Rat, Monkey