missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HDAC10 Recombinant Rabbit Monoclonal Antibody (23GB6165), Invitrogen™
GREENER_CHOICE

Product Code. p-200088295
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30089256 100 μL 100µL
30089593 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30089256 Supplier Invitrogen™ Supplier No. MA552992

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC10
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6165
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HDAC10
Gene Accession No. Q569C4, Q6P3E7, Q969S8
Gene Alias AW548891; DKFZp761B039; Hd10; Hdac10; Histone deacetylase 10; MGC149722; Polyamine deacetylase HDAC10; RP3-402G11.7
Gene Symbols HDAC10
Host Species Rabbit
Immunogen A synthesized peptide derived from human HDAC10 (345-383AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170787, 362981, 83933
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.