Learn More
HDAC11 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98764PCP
Product Code. 18699405
  • € 430.00 / 0.10mL
Description
HDAC11 Polyclonal antibody specifically detects HDAC11 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HDAC11
PerCP
Store at 4C in the dark.