missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC11 Recombinant Rabbit Monoclonal Antibody (23GB6005), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | HDAC11 |
|---|---|
| Clone | 23GB6005 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087573
![]() |
Invitrogen™
MA552970 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089802
|
Invitrogen™
MA552971 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC11 Recombinant Monoclonal antibody specifically detects HDAC11 in Human, Mouse samples. It is validated for Western BlotSpecifications
| HDAC11 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q91WA3, Q96DB2 | |
| 232232, 79885 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6005 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| FLJ22237; HD11; HDAC11; Histone deacetylase 11 | |
| A synthesized peptide derived from human HDAC11 (205-269AA). | |
| Primary | |
| -20°C | |
| HDAC11 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title