missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC2 Monoclonal Antibody (HDAC2-62)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA125451
This item is not returnable.
View return policy
Description
Recommended positive controls: NIH3T3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HDAC2 belongs to the histone deacetylase family. Histone deacetylases act via the formation of large multiprotein complexes and are responsible for the deacetylation of lysine residues at the N-terminal regions of core histones (H2A, H2B, H3 and H4). This protein forms transcriptional repressor complexes by associating with many different proteins, including YY1, a mammalian zinc-finger transcription factor. Thus, it plays an important role in transcriptional regulation, cell cycle progression and developmental events. Alternative splicing results in multiple transcript variants.
Specifications
| HDAC2 | |
| Monoclonal | |
| 2.5 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P56519, P70288, Q92769 | |
| HDAC2 | |
| Synthetic peptide corresponding to the C-terminal region of human histone deacetylase 2 (amino acid residues 471-488 with N-terminal added cysteine) conjugated to maleimide-activated KLH. | |
| 100 μL | |
| Primary | |
| Bovine, Canine, Chicken, Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| ChIP Assay, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| HDAC2-62 | |
| Unconjugated | |
| HDAC2 | |
| D10Wsu179e; HD2; hdac 2; Hdac2; hdac-2; Histone deacetylase 2; histone deacetylase 2-like protein; histone deacetylase-2; I79_010219; mRPD3; RPD3; transcriptional regulator homolog RPD3; YAF1; YY1 transcription factor-binding protein; YY1-associated factor 1; Yy1bp | |
| Mouse | |
| Purified | |
| RUO | |
| 15182, 3066, 395635, 407223, 475035, 84577 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction