missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585272
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Positive Control: 293T, A431, HeLa, HepG2, C2C12, Rat2, NB2a. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HDAC2 belongs to the histone deacetylase family. Histone deacetylases act via the formation of large multiprotein complexes and are responsible for the deacetylation of lysine residues at the N-terminal regions of core histones (H2A, H2B, H3 and H4). This protein forms transcriptional repressor complexes by associating with many different proteins, including YY1, a mammalian zinc-finger transcription factor. Thus, it plays an important role in transcriptional regulation, cell cycle progression and developmental events. Alternative splicing results in multiple transcript variants.
Specifications
| HDAC2 | |
| Polyclonal | |
| Unconjugated | |
| HDAC2 | |
| D10Wsu179e; HD2; hdac 2; Hdac2; hdac-2; Histone deacetylase 2; histone deacetylase 2-like protein; histone deacetylase-2; I79_010219; mRPD3; RPD3; transcriptional regulator homolog RPD3; YAF1; YY1 transcription factor-binding protein; YY1-associated factor 1; Yy1bp | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15182, 3066, 84577 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.16 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P70288, Q92769 | |
| HDAC2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human HDAC2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction