Learn More
Invitrogen™ HDAC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585378
Description
Keep as concentrated solution. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (89%), Pig (100%), Chicken (95%), Rhesus Monkey (100%), Bovine (100%). Positive Control: 293T, A431, HeLa, HepG2, U87-MG, SK-N-SH, IMR32, SK-N-AS, Neuro2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat2, NB2a. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. HDAC3 belongs to the histone deacetylase/acuc/apha family. It has histone deacetylase activity and represses transcription when tethered to a promoter. It may participate in the regulation of transcription through its binding with the zinc-finger transcription factor YY1. This protein can also down-regulate p53 function and thus modulate cell growth and apoptosis. This gene is regarded as a potential tumor suppressor gene.
Specifications
| HDAC3 | |
| Polyclonal | |
| Unconjugated | |
| HDAC3 | |
| AW537363; HD3; Hdac3; HDAC3 splicing HDAC3alpha; HDAC3 splicing HDAC3beta; HDAC3 splicing HDAC3delta; HDAC3 splicing HDAC3gamma; histone deacetylase 3; histone deacetylase 3-like protein; RPD3; RPD3-2; SMAP45 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15183, 84578, 8841 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.32 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O15379, O88895, Q6P6W3 | |
| HDAC3 | |
| Recombinant protein encompassing a sequence within the center region of human HDAC3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.