missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78981AF350
This item is not returnable.
View return policy
Description
HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| HDAC6 | |
| Polyclonal | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
| Rabbit | |
| 0.1 mL | |
| 10013 | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HDAC6 | |
| A synthetic peptide made to a C-terminal portion of the human HDAC6 protein (between residues 1175-1215). [UniProt Q9UBN7] | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction