missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41286AF750
This item is not returnable.
View return policy
Description
HDAC6 Polyclonal antibody specifically detects HDAC6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| HDAC6 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
| Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human HDAC6. The immunogen is located within the last 50 amino acids of HDAC6. Amino Acid Squence: NIAHQNKFGEDMPHPH | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Neuroscience | |
| 10013 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction