missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61065
This item is not returnable.
View return policy
Description
HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| HDAC6 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2 - 10 ug/mg lysate | |
| NP_006035.2 | |
| HDAC6 | |
| The immunogen recognized by this antibody maps to a region between residue 1165 and 1215 of human histone deacetylase 6 using the numbering given in entry NP_006035.2 (GeneID 10013). | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases | |
| 10013 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction