missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA511240
This item is not returnable.
View return policy
Description
HDAC6 Polyclonal Antibody for Western Blot, IHC (P)
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.
Specifications
| HDAC6 | |
| Polyclonal | |
| Unconjugated | |
| HDAC6 | |
| CPBHM; FLJ16239; Hd6; HDA6; Hdac5; Hdac6; histone deacetylase 5; histone deacetylase 6; Histone deacetylase mHDA2; JM21; KIAA0901; mHDA2; PPP1R90; protein phosphatase 1, regulatory subunit 90; scurfy candidate 6; Sfc6 | |
| Rabbit | |
| Protein A, Affinity chromatography, Antigen affinity chromatography | |
| RUO | |
| 10013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9UBN7 | |
| HDAC6 | |
| KLH conjugated synthetic peptide between 1182-1215 amino acids from the C-terminal region of human HDAC6. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction