missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HDAC6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16586441
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16586441 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16586441 Supplier Invitrogen™ Supplier No. PA588129

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: NEGELRVLAD TYDSVYLHPN SYSCACLASG SVLRLVDAVL GAEIRNGMAI IRPPGHHAQH SLMDGYCMFN HVAVAARYAQ QKHRIRRVLI VDWDVHHGQG TQFTFDQDPS VLYFSIHRYE QGRFWPHLKA SNWSTTGFGQ GQGYTINVPW NQVGMRDADY IAAFLHVLLP VALEFQPQLV LVAAGFDALQ GDPKGEMAAT PAGFAQLTHL LMGLAGGKLI LSLEGGYNLR ALAEGVSASL HTLLGDPCPM LESPGAPCRS AQASVSCALE ALEPFWEVLV RSTETVERDN MEEDNVEESE EEGPWEPPVL P.

Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC6
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HDAC6
Gene Accession No. Q9UBN7, Q9Z2V5
Gene Alias CPBHM; FLJ16239; Hd6; HDA6; Hdac5; Hdac6; histone deacetylase 5; histone deacetylase 6; Histone deacetylase mHDA2; JM21; KIAA0901; mHDA2; PPP1R90; protein phosphatase 1, regulatory subunit 90; scurfy candidate 6; Sfc6
Gene Symbols HDAC6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-470 of human HDAC6 (NP_0060352).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10013, 15185
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.