missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98759-50ul
This item is not returnable.
View return policy
Description
HDAC7 Polyclonal antibody specifically detects HDAC7 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| HDAC7 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunoprecipitation 1-2 μL/mg of lysate | |
| DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HDAC7. | |
| 50 μg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics | |
| 51564 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction