missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HDAC7 Antibody (PCRP-HDAC7-1B6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26705-100ug
This item is not returnable.
View return policy
Description
HDAC7 Monoclonal antibody specifically detects HDAC7 in Human samples. It is validated for Flow Cytometry
Specifications
| HDAC7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-HDAC7-1B6 | |
| Unconjugated | |
| DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917 | |
| Recombinant full-length human HDAC7 protein | |
| 100 μg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Phospho Specific | |
| 51564 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction