missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC7 Recombinant Rabbit Monoclonal Antibody (6J2N9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535176
This item is not returnable.
View return policy
Description
Immunogen sequence: RPQRLHHHLF LAGLQQQRSV EPMRLSMDTP MPELQVGPQE QELRQLLHKD KSKRSAVASS VVKQKLAEVI LKKQQAALER TVHPNSPGIP YRTLEPLETE GATRSMLSSF LPPVPSLPSD PPEHFPLRKT VSEPNLKLRY KPKKSLERRK NPLLRKESAP PSLRRRPAET LGDSS.
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene has sequence homology to members of the histone deacetylase family. This gene is orthologous to mouse HDAC7 gene whose protein promotes repression mediated via the transcriptional corepressor SMRT. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| HDAC7 | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8C2B3, Q8WUI4, Q99P96 | |
| HDAC7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 24-198 of human HDAC7 (Q8WUI4). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 6J2N9 | |
| Unconjugated | |
| HDAC7 | |
| 5830434K02Rik; HD7; HD7a; hdac4l; Hdac7; hdac-7; Hdac7a; histone deacetylase 7; Histone deacetylase 7A; mFLJ00062 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51564, 56233, 84582 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction