missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC9 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-03993UV
This item is not returnable.
View return policy
Description
HDAC9 Polyclonal specifically detects HDAC9 in Human, Mouse, Bovine, Canine, Primate samples. It is validated for Western Blot.
Specifications
| HDAC9 | |
| Polyclonal | |
| Western Blot | |
| EC 3.5.1.98, HD7b, HD7HDAC, HD9, HDAC7, HDAC7BDKFZp779K1053, HDRP, histone deacetylase 4/5-related protein, Histone deacetylase 7B, histone deacetylase 9, Histone deacetylase-related protein, KIAA0744HDAC9B, MEF-2 interacting transcription repressor (MITR) protein, MEF2-interacting transcription repressor MITR, MITRHDAC9FL | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HDAC9 | |
| A portion of amino acids 100-150 of human HDAC-9 was used as the immunogen. | |
| 0.1 mL | |
| Cell Cycle and Replication, Epigenetics | |
| Human, Mouse, Bovine, Canine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction