missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDC Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598315
This item is not returnable.
View return policy
Description
HDC Polyclonal Antibody for Western Blot, IHC (P), ELISA
The biogenic amine histamine is an important modulator of numerous physiologic processes, including neurotransmission, gastric acid secretion, and smooth muscle tone. The biosynthesis of histamine from histidine is catalyzed by the enzyme L-histidine decarboxylase (HDC; EC 4.1.1.22). This homodimeric enzyme is a pyridoxal phosphate (PLP)-dependent decarboxylase and is highly specific for its histidine substrate.
Specifications
| HDC | |
| Polyclonal | |
| Unconjugated | |
| Hdc | |
| AW108189; HDC; Hdc-a; Hdc-c; Hdc-e; Hdc-s; histidine decarboxylase; L-histidine decarboxylase; MGC163399 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 3067 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.51 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P19113 | |
| Hdc | |
| Recombinant Human Histidine decarboxylase protein (504-648AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction