missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HDG11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17189485 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17189485 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17189485 Supplier Invitrogen™ Supplier No. PA5114314

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody specifically recognizes HDG11 in Arabidopsis thaliana.

Transcription factor which acts as positive regulator of drought stress tolerance. Can transactivate CIPK3, NCED3 and ERECTA. Transactivates several cell-wall-loosening protein genes by directly binding to HD motifs in their promoters. These target genes play important roles in coordinating cell-wall extensibility with root development and growth. Transactivates CYP74A/AOS, AOC3, OPR3 and 4CLL5/OPCL1 genes by directly binding to HD motifs in their promoters. These target genes are involved in jasmonate (JA) biosynthesis, and JA signaling affects root architecture by activating auxin signaling, which promotes lateral root formation. Acts as negative regulator of trichome branching . Required for the establishment of giant cell identity on the abaxial side of sepals . May regulate cell differentiation and proliferation during root and shoot meristem development. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDG11
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene HDG11
Gene Accession No. Q9FX31
Gene Alias AT1G73360; AtEDT1; ATHDG11; EDT1; ENHANCED DROUGHT TOLERANCE 1; HDG11; HDGL2-11; HD-ZIP protein HDG11; Homeobox-leucine zipper protein HDG11; Homeodomain GLABRA 2-like protein 11; homeodomain GLABROUS 11; Homeodomain transcription factor HDG11; Protein ENHANCED DROUGHT TOLERANCE 1; Protein HOMEODOMAIN GLABROUS 11; putative homeobox protein; T9L24.43; T9L24_43
Gene Symbols HDG11
Host Species Rabbit
Immunogen Synthetic peptide within thaliana HDG11 aa 631-680.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 843671
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.